Would you like to log in again?
MixMate® Mixer
Supplier: Eppendorf
MixMate®
5353000529 5353000529
76420-660EA 2550.28 USD InStock
76420-660 CA76420-660
MixMate® Mixer
Shakers and Mixers Microplate Shakers
Mix samples completely and reliably with the Eppendorf® MixMate. 96 and 384-well plates and reaction tubes are mixed by the 2DMix-Control technology even at the microliter scale. The additional vortex function has a 2 s run timer allowing processing your next sample without restarting.
  • High-speed mixer (3000 rpm) for fast and efficient mixing of small volumes in all common vessel formats up to 50 ml
  • 2DMix-Control technology (planar mixing orbit without vertical movement) for controlled mixing and reproducible results in seconds
  • Anti-spill technology for controlled mixing movements without lid wetting and cross-contamination
  • Five different tube holders (0.5 ml, 1.5/2.0 ml, 5/15 ml, 30/50 ml, PCR 96-well plates) for high vessel flexibility
  • Integrated ergonomic vortex function for various vessel formats for distinct sample mixing
  • Minimal hand/arm vibration transfer during vortexing (certificate available) for ergonomic operation (PhysioCare Concept®)
  • Five soft keys with pre-defined standard mixing parameters for easy usage
  • Very stable remains stationary even at maximum mixing speed
  • Whisper quiet operation to benefit the work environment
Mix samples in seconds, fully and reliably. Whether in plates (96 or 384-wells) or reaction tubes, your samples will be optimally processed. With the integrated ergonomic vortex function, the MixMate is your perfect laboratory assistant.
Other functions include 2D Mix-control, anti-spill technology, touch vortexing.
Mixing and vortexing radius: 1.5 mm
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty