Would you like to log in again?
Sterisart® NF Sterility Test Systems, Sartorius
Supplier: Sartorius
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
16477GBD 16468GBD 16467GBD 16475GBD 16420 16419 16475GSD 16478GBD 16419 16466GBD 16469GBD 16420 16470GBD 16469GSD 16476GSD 16476GBD 16466GSD 16467GSD
76420-838PK 632.67 USD InStock
76420-838 76420-816 76420-826 76420-814 76420-836 76420-824 76409-140 76420-828 76409-142 CA76409-140 76420-830 76420-840 CA76409-142 76420-834 76420-822 76420-832 76420-820 76420-842
Sterisart® NF Sterility Test Systems, Sartorius
Filters Membrane Filter Units
The Sterisart® NF System is a completely disposable technology based on the membrane filtration method, for the sterility testing of pharmaceutical products. It makes aseptic sampling easy, during or after incubation which is required for dilution or rapid detection, or for the supplement of additives for inactivation of antibiotics.
  • Sterisart NF alpha, dual-needle metal spike for closed containers
  • Sharp Metal needle for easy penetration
  • High retention of microbes
  • Low adsorption
  • Huge variety of adapters and more than 20 different unit types
  • Gas-impermeable packaging for protection against sterilant
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty