Would you like to log in again?
SANDWASH MASK WITH EAR LOOPS CS500
Supplier: VWR International
Sandwash mask^ Sandwash mask, features two layers of Sandwash fabric, latex-free elastic ear loops, and a form-fitting nose bridge, Typically used for cleanroom garments because of it's tight weave and low linting properties, Sandwash masks will not shrink or stretch providing consistant filtration
ORDER
Expand All / Collapse All
Description VWR Catalog Number Unit Price Quantity
SANDWASH MASK WITH EAR LOOPS CS500 76420-868 Case of 500 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty