Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

MAbPac™ RP LC Columns

Supplier: Thermo Scientific
MAbPac™ RP
303183 088648 303269 303270 088645 088647 303184 088646 088649 303270 088644
76424-342EA 1237.56 USD InStock
76424-342 10783-114 10783-112 76420-870 10783-108 76424-344 10783-106 10783-116 76420-874 76320-034 10783-110
MAbPac™ RP LC Columns
Chromatography Columns
MAbPac RP is a reverse phase (RP) column specifically designed for separation of intact monoclonal antibodies (mAbs) and mAb fragments.

  • Superior resolution power for monoclonal antibodies and related substances
  • Ideal for small precious samples
  • High efficiency with low carry-over
  • For direct injection into MS: 75 to 150 µl/min
  • Wide operating pH range
  • High temperature stability, up to 110 °C
  • Optimized for low flow rates

The stationary phase is fully compatible with mass spectrometry friendly organic solvent such as acetonitrile and isopropanol, as well as low pH eluents containing trifluoroacetic acid or formic acid.

The MAbPac RP is based on wide-pore 4 µm polymer particles that are stable at extreme pH (0 to 14) and high temperature (up to 110 °C). The wide- pore size of polymeric particles enables efficient separation of protein molecules with low carry-over.

This column can be used to separate intact mAb and variants, light chain (LC) and heavy chain (HC), Fc and Fab fragments, scFc and F(ab’)2 fragments , using LC/UV or LC/MS.

Ordering information: Guard cartridges require use of the corresponding guard holder. See Acclaim Guard Kit Holder and Coupler V-2.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty