Would you like to log in again?
CX-1 PH GRADIENT BUFFER B PH 10.2 1L
Supplier: Thermo Scientific
CX-1 pH Gradient Buffer B^ CX-1 pH Gradient Buffer B, pH 10.2, Deliver robust, reproducible gradients with the pH buffer platform, which is applicable to a wide range of MAbs and ready to use with existing LC columns and systems, without the need to formulate mobile phases to QA/QC, Size: 1l
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
CX-1 PH GRADIENT BUFFER B PH 10.2 1L 76420-884 303275 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty