Would you like to log in again?
C4 PREPARATIVE COLUMN 5UM 250X21.2MM
Supplier: Thermo Scientific
C4 preparative column^ C4 preparative column, Particle size: 5um, Attain faster separations for hydrophobic analytes when you scale up use C4 reversed phase preparative HPLC Column, for your purification need, provide excellent sensitivity and high efficiency, lower hydrophobicity, Size: 250X21.2mm
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
C4 PREPARATIVE COLUMN 5UM 250X21.2MM 76420-888 72305-259270A Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty