Would you like to log in again?
PFP HPLC COLUMN 5 UM PORE SIZE 250X21MM
Supplier: Thermo Scientific
PFP HPLC Column^ PFP HPLC Column, Use for analyzing difficult-to-resolve mixtures of halogenated compounds including pesticides, Particle size: 5 um, Ph: 5 um, Surface Area: 220 m2/g, Silica, Spherical Porous Ultrapure, Excellent peak symmetry, Narrow peaks for outstanding efficiency, Size: 250X21mm
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
PFP HPLC COLUMN 5 UM PORE SIZE 250X21MM 76420-890 25405-259270A Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty