Would you like to log in again?
PolarSafe® Cooling Transport Bags, Cole Parmer
Supplier: Cole Parmer
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
PolarSafe®
03726-02 03726-39 03726-03 03726-35 03726-38 03726-34 03726-32 03726-36 03726-33 03726-37 03726-01 03726-00
76420-948EA 743.44 USD InStock
76420-948 76420-926 76420-946 76420-928 76420-940 76420-950 76420-944 76420-954 76420-932 76420-942 76420-930 76420-952
PolarSafe® Cooling Transport Bags, Cole Parmer
Boxes Cooling/Freezing Boxes and Bags
These insulated nylon bags provide an easy solution for transporting your samples.
  • Maintain specific temperature ranges for extended periods of time for improved sample protection
  • Get the temperature protection you need with choice of durable transport bags or boxes
  • Modular, color-coded cooling packs allow flexible configurations for different-sized samples
  • Ensure sample integrity with optional security sealing ties
Organize cooling packs to suit your sample or choose from one of our prepared kits. Bags have pockets to keep documentation safe during transit, a shoulder strap, and carrying handles.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty