Would you like to log in again?
PolarSafe® Dewar and Traceable® Temperature Data Logger Bundles, Cole Parmer
Supplier: Cole Parmer
PolarSafe® PolarSafe™
44260-75 44260-72 44260-74 44260-73 44260-76 44260-71
76421-000EA 9947.04 USD InStock
76421-000 76421-010 76421-002 76421-004 76421-006 76421-008
PolarSafe® Dewar and Traceable® Temperature Data Logger Bundles, Cole Parmer
Dewars and Cryogenic Vessels
The Cole-Parmer® PolarSafe® dewar together with a Traceable® data logger is the perfect combination to safely store your critical samples while maintaining sample integrity.
  • Improve sample protection – maintain specific temperature ranges for extended time periods
  • Maintain sample integrity with Traceable® temperature monitoring
  • Data logger provided with 2-year NIST-traceable calibration certificate
  • Lowest set temperature of –200 °C
  • Durable aluminum construction
The bundle adds on one Traceable® Wi-Fi data logger compatible with TraceableLIVE® cloud service (model 18001-08) designed specifically for temperatures down to –200 °C. Once your data logger is set up, you can securely monitor parameters 24/7, control up to 100 alarms remotely, view your data logging history, run reports in real time, and give access to other team members.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty