Would you like to log in again?
TOTAL RNA LIBRARY KIT FASTEST 96 PREPS
Supplier: Zymo Research
Seq RiboFree Total RNA Library Kit^ Seq RiboFree Total RNA Library Kit, is the fastest, easiest RNA-Seq library prep kit available to stranded, total RNA libraries, automation-friendly protocol minimizes hands-on time to generate sequencer-ready, indexed libraries depleted of rRNA, Size: 96preps
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
TOTAL RNA LIBRARY KIT FASTEST 96 PREPS 76421-012 R3003 Each Retrieving
Temperature sensitive Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty