Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
SEQUENCE UDI PRIMER SET UNIQUE 96INDEXES
Supplier: Zymo Research
Sequence UDI Primer Set^ Seuence UDI Primer Set, contain pre-mixed Unique Dual Index (UDI) primers that can be used to barcode any library containing TruSeq adapters, Indexes in the Seq UDI Primer Set (Indexes 1-12) are dispensed in 1.5 mL tubes, and  -Seq UDI Primer Plate, Size: 96 indexes
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
SEQUENCE UDI PRIMER SET UNIQUE 96INDEXES 76421-014 D3096 Each (10µl) Retrieving
Temperature sensitive Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty