Would you like to log in again?
GUT MICROBIOME STANDARD 2X DNA 10 PREPS
Supplier: Zymo Research
Gut Microbiome Standard^ Gut Microbiome Standard, 2X DNA shield, is a an accurately quantified microbial community mimicking the human gut microbiome, The challenges it presents to microbiomics profiling researchers to validate protocols, Impurity level: < 0.01% foreign microbial DNA, Size: 10 preps
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
GUT MICROBIOME STANDARD 2X DNA 10 PREPS 76421-016 D6331 Each (750µl) Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty