Would you like to log in again?
BenchBlotter Platform Rocker and Accessories, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
65700-01
76421-776EA 964.89 USD InStock
76421-776
BenchBlotter Platform Rocker and Accessories, Electron Microscopy Sciences
Shakers and Mixers Rotating/Rocking Shakers
The BenchBlotter 2D Rocker is made for common molecular lab applications.
  • Easy-to-use
The BenchBlotter 2D Rocker is lightweight and can be easily transported to incubators or cold rooms. Optional stacking platforms will double your work area. The speed is fixed at 12 rpm for 115 V and 10 rpm for the 230 V. Racks provide 3" separation.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty