Would you like to log in again?
BenchRocker™ 2D Variable Speed Rocker, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
65700-06
76421-786EA 1507.87 USD InStock
76421-786
BenchRocker™ 2D Variable Speed Rocker, Electron Microscopy Sciences
Shakers and Mixers Rotating/Rocking Shakers
The BenchRocker™ 2D Rocker used for many molecular and biological mixing applications.
  • Large 14×12" work surface
The BenchRocker™ 2D Rocker provides the exact speeds and tilt angles needed for mixing applications. Set the parameters to match your vessel size and volume of liquid for best results. It features a large work surface and is safe for incubators and cold rooms. The optional stacking platform gives you twice the work area.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty