Would you like to log in again?
Tube Revolver, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
71785
76421-798EA 761.66 USD InStock
76421-798
Tube Revolver, Electron Microscopy Sciences
Shakers and Mixers Rotating/Rocking Shakers
Using a combination of up to four paddle the tube revolver can hold a combination of paddles.
  • Maintenance-free motor with quiet operation
  • Plug and Play operation
  • Adjustable paddle angle
  • Includes five interchangeable rotisseries
With seven 10/15 ml tubes, seven 5/7 ml tubes, fifteen 1.5/2.0 ml tubes, or twenty one 0.5/0.8 ml tubes. Or one paddle of six 50 ml tubes.
Max. power: 10 W
Delivery information: Includes 2 each paddles with for 10/15 ml tubes (7 tubes/paddle), 5/7 ml tubes (7 tubes/paddle), 1.5/2 ml tubes (17 tubes/paddle), 0.5/0.8 ml tubes (21 tubes/paddle), or 50 ml tubes (6 tubes/paddle).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty