Would you like to log in again?
Table-Gard Disposable Work Mats, Beach Waves, Accuform
Supplier: Accuform
ACCUFORM®
LPM104X LPM104P LPM104L
76421-884PK 18.74 USD InStock
76421-884 76421-886 76421-888
Table-Gard Disposable Work Mats, Beach Waves, Accuform
Surface Protectors Bench Protectors
Create safe, clean workspaces by applying the adhesive mat to surfaces that are not regularly cleaned.
  • Provides protection from germs, dirt, and chemical residue
  • Compact and lightweight mat fits in workbags for on-the-go working needs
  • Made of a smooth, thin 2-mil plastic film that is waterproof and tear-resistant
  • Can be discarded in waste or recycle bin (recycle code 5)
  • 12.5×19.5" size creates a workspace large enough for laptops and folios
  • BPA, Latex, and PVC-free material without toxic chemicals
  • The adhesive is safe for indirect food contact (complies with FDA 21 CFR 175.105)
The self-adhesive mat is repositionable and lifts easily for disposal when work is complete. The mat is safe for application to countertops, desktops, conference tables, and restaurant tables.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty