Would you like to log in again?
BAG 95KPA PRESSURE TESTED CS500
Supplier: COM-PAC INTERNATIONAL INC.
Bag^ Bag, 95kPa pressure tested, feature leak-proof side seals and tamper-evident adhesive closure, Specimen tubes and absorbent materials can be securely sealed within the front pocket of the bag by peeling away the paper liner and folding the adhesive down onto itself, forming pressure-tight seal
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
BAG 95KPA PRESSURE TESTED CS500 76422-046 XP-95-0609 Case of 500 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty