Would you like to log in again?
Sterile SatPax® 550 Pre-Wetted 100% Meltblown Polypropylene Cleanroom Wiper, Berkshire
Supplier: Berkshire
SSP55000424 SSP55000524
76422-916CS 812.09 USD InStock
76422-916 76097-918
Sterile SatPax® 550 Pre-Wetted 100% Meltblown Polypropylene Cleanroom Wiper, Berkshire
Wipes Cleanroom Wipes Presaturated Cleanroom Wipes
Sterile SatPax® 550 wipers are composed of exceptionally clean, 100% meltblown polypropylene.
  • Designed for use in ISO Class 5 and higher cleanroom environments
  • Very low extractables and ions
  • Very low particle and fiber counts
  • Reduces alcohol usage and preparation/handling costs
  • Reduces VOC emissions
  • Increases cleaning efficiency
  • Increases cleaning protocol consistency
  • Resealable solvent-resistant packaging
Sterile SatPax® 550 wipers are composed of exceptionally clean, 100% meltblown polypropylene, pre-wetted with 70% IPA and 30% DI water. This pre-wetted format provides a cost effective and easy to use solution versus traditional bulk handling of solvents, maintenance of squirt bottles, and inconsistent wetting and cleaning associated with wetting a dry wiper. Each package is gamma irradiated sterile to a 10⁻⁶ Sterility Assurance Level, making it ideal for cleaning critical sterile processing environments.
Certifications: Gamma irradiated and sterile validated to a 10⁻⁶ Sterility Assurance Level per AAMI guidelines. ISO Class 5 – 8, Cleanroom Class 100 – 100,000, EU Grade A/B/C/D. EC Regulation 1935 (2004)- Food contact compliant. FDA 21 CFR-Food contact compliant.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty