Would you like to log in again?
CHAIR BTA ESD W/GLIDES VINYL BL 19-26IN
Supplier: BIOFIT ENGINEERED SEATING
Chair^ Bridgeport series Chair, ESD static control rated, Features: 18.5in W x 16.5in D x 3in T seat, 14.5in W x 9.5 H backrest with lumbar support, 25.6in diameter cast aluminum base, 19-26in seat height range, chrome metal finish, glides, blue vinyl upholstery
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
CHAIR BTA ESD W/GLIDES VINYL BL 19-26IN 76422-964 BTAMHTXXKCMUA Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty