Would you like to log in again?
BioFit Bridgeport Cleanroom ESD Chairs, ISO 5 ESD
Supplier: BioFit
Bridgeport
BTAMRTFX5KCMUA BTSHHTXX5KCMUA BTALHTXX5KCMUA BTSLRTXX5KCMUA BTSMRTXX5KCMUA BTSHRTXX5KCMUA BTAHRTPX5KCMUA BTSMHTXX5KCMUA BTALRTXX5KCMUA BTAMHTFX5KCMUA BTAMHTXX5KCMUA
76422-986EA 1627.64 USD InStock
76422-986 76422-996 76423-014 76423-006 76422-972 76422-950 76422-982 76423-010 76423-012 76422-966 76422-954
BioFit Bridgeport Cleanroom ESD Chairs, ISO 5 ESD
Chairs
Bridgeport series chairs boast a number of features and benefits that make them a popular choice for laboratory, healthcare, education and technical workspaces. The ergonomic design enhances user comfort and productivity when sitting for prolonged periods.
  • ISO 5 Cleanroom ESD combination chair
  • Easily adjust seat-height from seated position
  • Height and tilt adjustable backrests with lumbar support
  • Dual-wheel resistance casters enhance user safety by inhibiting seating from rolling away on hard floors or 2"" high polyamide glides
  • Adjustable and fixed height footrings
Seating solutions are crafted with user comfort, health and wellbeing in mind. Products meet or exceed rigorous industry standards and nearly all carry a 13-year warranty with a lifetime warranty on the pneumatic mechanisms.
The products are renowned for proven performance and reliability, backed by a history of over 75 years improving the workplace experience by providing ergonomic seating for laboratory, healthcare, technology, education, industrial, office and custom applications.
Certifications: LEED® compliant for use in commercial interiors complies with ANSI/BIFMA X5.1 product safety and performance standards.
Caution: This product can expose you to chemicals including wood dust, which is known to the State of California to cause cancer. For more information go to www.p65warnings.ca.gov.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty