Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

QuantiChrom™ Carbonyl Assay Kit, BioAssay Systems

Supplier: BioAssay Systems
DCAR-100
76423-318EA 416.95 USD InStock
76423-318
QuantiChrom™ Carbonyl Assay Kit, BioAssay Systems
Assays Protein Assays
Quantification of carbonyl groups or protein carbonyls in biological samples.

  • Fast and sensitive: use of 100 µl sample, linear detection range from 12 to 250 µm carbonyl in 96-well plate assay
  • The procedure involves adding a single working reagent, and reading the absorbance after 30 minutes
  • Detection limit: 12 µm
  • Method: OD375nm
  • Samples: chemicals with carbonyl groups or protein carbonyls in biological samples

Carbonyl groups, such as ketones and aldehydes, are a commonindicator of protein oxidation. Protein oxidation is caused as a result of exposure to reactive oxygen species (ROS) and is a common marker in various diseases as well as aging. BioAssay Systems' carbonyl assay kit is based on an improved method, where 2,4-dinitrophenylhydrazine (DNPH) reacts with carbonyl groups to produce a colored compound at 375 nm. The intensity of this colored compound is directly proportional to the carbonyl groups in the sample.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty