Would you like to log in again?
QuantiChrom™ Boron Assay Kit, BioAssay Systems
Supplier: BioAssay Systems
DBOR-100
76423-328EA 405.32 USD InStock
76423-328
QuantiChrom™ Boron Assay Kit, BioAssay Systems
Assays Protein Assays
Quantitative determination of boron in agricultural and environmental samples.
  • Fast and sensitive: linear detection range: 0.05 to 10 µg/ml (0.05 to 10 ppm) boron with 100 µl sample (96-well)
  • Method: OD 420 nm
  • Samples: water, plant tisse, soil samples, and antibody conjugation solutions
Boron is an essential micronutrient in plants and is involved in maintaining robust cell walls, cell membranes, and reproductive tissues. Although boron is common in the soil in its natural state as a borate mineral, the amount of boron available to plants is actually quite small. As a result, boron deficiency is the second most common micronutrient deficiency among crop plants. In order to keep plant boron levels in a healthy range, supplementation to the soil via fertilizers and additives is often required. If not regulated, a lack of or excess of boron may significantly lower crop yield. In the biotech industry, sodium borohydride is commonly used to conjugate antibodies and typically needs to be removed from the final product, especially for therapeutic antibodies.
BioAssay Systems boron detection kit provides a convenient and reliable means to measure boron. In the assay borate complexes with azomethine-H to create a colored compound that can be measured at 420 nm. This assay can be used with a variety of samples and is simple, sensitive, and adaptable to high-throughput screening.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty