Would you like to log in again?
QuantiChrom™ Hyaluronidase Inhibitor Screening Assay Kit, BioAssay Systems
Supplier: BioAssay Systems
DIHY-100
76423-336EA 575.1 USD InStock
76423-336
QuantiChrom™ Hyaluronidase Inhibitor Screening Assay Kit, BioAssay Systems
Assays Enzyme Assays
High-throughput screening for evaluation of hyaluronidase inhibitors.
  • Rapid and reliable: entire assay can be completed in 30 minutes
  • Method: turbidimetric (OD 600 nm)
  • Samples: test compounds, hyaluronidase inhibitors
Hyaluronidases are a family of enzymes that catalyze the degradation of the glycosaminoglycan, hyaluronic acid. Hyaluronic acid is one of the major constituents of the extracellular matrix in organisms where it contributes to both cell proliferation and migration. The role of hyaluronidases in breaking down this key factor in cell proliferation makes them a possible target for cancer treatment. One hypothesis is that increased hyaluronidase may help prevent tumor invasion by breaking down the extracellular matrix needed for tumor expansion. Conversely, decreasing hyaluronidase activity might prevent metastasis by stopping cancer cells from escaping primary tumor masses. The study to determine hyaluronidases exact role in cancer pathology is still ongoing.
BioAssay Systems hyaluronidase inhibitor screening assay kit uses a two step turbidimetric reaction to measure hyaluronidase activity by the amount of hyaluronic acid that is hydrolyzed. A stop reagent halts the enzymatic reaction and forms turbidity with any residual hyaluronic acid in the well. The decrease in turbidity at 600 nm is directly proportional to hyaluronidase activity in the sample.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty