Would you like to log in again?
NEBExpress MBP Fusion and Purification System, New England Biolabs
Supplier: New England Biolabs
E8201S
76423-408EA 800 USD InStock
76423-408
NEBExpress MBP Fusion and Purification System, New England Biolabs
Protein Purification Protein Purification Kits
The NEBExpress MBP fusion and purification system takes advantage of the strong Ptac promoter and the translation initiation signals of maltose binding protein (MBP) to enhance solubility and expression levels of a desired protein in E. coli. The resulting product is an MBP fusion protein, which is then purified by affinity chromatography.
  • Reliable E. coli expression: substantial yields (up to 100 mg/L)
  • Fusion to MBP has been shown to enhance the solubility of proteins expressed in E. coli
  • Two-step purification: amylose elution followed by TEV Protease cleavage and Ni resin isolation
  • Gentle elution with maltose; no detergents or harsh denaturants required
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty