Would you like to log in again?
Tth Argonaute (TtAgo)
Supplier: New England Biolabs
M0665S
76423-412EA 83 USD InStock
76423-412
Tth Argonaute (TtAgo)
Enzymes
TtAgo is a prokaryotic argonaute from the Gram-negative, thermophilic bacterium Thermus thermophilus which functions as a DNA-guided endonuclease when provided with a 16-18 nucleotide long 5-phosphorylated single-stranded DNA oligonucleotide guide.
  • Short 16-18 ssDNA guides are cost effective; can be phosphorylated with T4 PNK
  • Guide/target sequence selection is not limited by the requirement of an adjacent sequence motif
  • Highly active on ssDNA and most dsDNA substrates (generates a nick in dsDNA substrates)
  • Mild activity on ssRNA substrates
  • Recommended for in vitro applications
The RNase H-like active site of TtAgo requires divalent Mg²⁺ (or Mn²⁺) metal ions for the reaction to occur and cleaves a complementary substrate between the bases corresponding to positions 10 and 11 of the DNA guide.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty