Would you like to log in again?
pMAL-c6T Vector, New England Biolabs
Supplier: New England Biolabs
N0378S
76423-414EA 160 USD InStock
76423-414
pMAL-c6T Vector, New England Biolabs
Vectors, Plasmids and Libraries
The pMAL-c6T Vector is designed to produce maltose-binding protein (MBP) fusions in the cytoplasm.
  • pMAL-c6T is the recommended vector for use with the NEBExpress MBP fusion and purification system
  • Fusion to MBP significantly enhances the solubility and proper folding of proteins in E. coli
  • Includes TEV Protease site, allowing protein to be cleaved from MBP using TEV Protease (NEB #P8112)
  • Two-step purification: amylose elution followed by TEV Protease cleavage and Ni resin isolation
The MBP has been engineered for tighter binding to amylose resin. The vector expresses an N-terminal his-tagged mal E gene followed by a multiple cloning site containing a TEV protease recognition sequence, allowing MBP to be cleaved from the protein of interest after purification.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty