Would you like to log in again?
Minimal M9 Broth Bottled Media, Sterile, Teknova
Supplier: Teknova
M8000 M8000
76423-596EA 79.77 USD InStock
76423-596 CA100219-260
Minimal M9 Broth Bottled Media, Sterile, Teknova
Media for Microbiology Prepared Media for Microbiology
For the maintenance and propagation of E.coli.
  • 1 Liter
  • Sterile
Contains 1× M9 salts (42 mM Na₂HPO₄, 24 mM KH₂PO₄, 9 mM NaCl, 19 mM NH₄Cl), 1 mM MgSO₄, 0.1 mM CaCl₂, 2.0% glucose, 0.5 µg/ml thiamine. Ambient shipping conditions. Store at room temperature.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty