Would you like to log in again?
Terrific Broth Complete with Animal-Free Soytone Bottled Media, Sterile, Teknova
Supplier: Teknova
T7660 T7660
76423-620EA 61.07 USD InStock
76423-620 CA100219-868
Terrific Broth Complete with Animal-Free Soytone Bottled Media, Sterile, Teknova
Media for Microbiology Prepared Media for Microbiology
For the growth of recombinant E.coli
  • No animal-based components
  • Sterile
Due to the enriched formulation of TB, strains have an extended growth phase and this results in high cell yields. TB is commonly used for plasmid purification and protein production. Ingredients: 2% animal-free soytone (20 g/L), 2.4% yeast xxtract (24.0 g/L), 0.4% glycerol (4.0 g/L), 72 mM dipotassium phosphate, 17 mM mono potassium phosphate. Ambient shipping conditions. Store at room temperature.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty