Would you like to log in again?
2YT Broth Bottled Media, Teknova
Supplier: TEKNOVA INC
Y0166 Y0166 Y0167
76423-630EA 57.16 USD InStock
76423-630 CA100219-100 CA100220-314
2YT Broth Bottled Media, Teknova
Media for Microbiology Prepared Media for Microbiology
2YT broth.
  • 500 ml and 1 Liter
  • Sterile
Contains 1.6% tryptone, 1.0% yeast extract, 0.5% NaCl. Ambient shipping conditions. Store at room temperature.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty