Would you like to log in again?
YPD Broth Bottled Media, Teknova
Supplier: Teknova
Y5000 Y5000
76423-632EA 86.81 USD InStock
76423-632 CA100219-830
YPD Broth Bottled Media, Teknova
Media for Microbiology Prepared Media for Microbiology
YPD is a nutritionally-rich, undefined, non-selective media for the growth and maintenance of Saccharomyces cerevisiae.
  • 1 Liter
  • Sterile
Contains 1.0% yeast extract, 2.0% peptone, 2.0% glucose. Ambient shipping conditions. Store at room temperature.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty