Would you like to log in again?
Corning® 3D Clear Antibody Buffer, Corning
Supplier: Corning
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Corning®
5735 5734
76423-686CS 103.41 USD InStock
76423-686 76423-688
Corning® 3D Clear Antibody Buffer, Corning
Buffers Cell Culture Buffers
Corning 3D Clear Antibody Buffer used with immunolabeling in Corning 3D tissue labeling approach.
  • Rapid tissue clearing
  • Easy to Use
  • No Special Equipment Required
  • Compatible with IF, FP and other Fluorescent lables
  • Microplate and Automation Compatible
  • Reversible to Follow up with 2D H&E/IHC
The Corning 3D Clear Antibody Buffer is used with immunolabeling in the Corning 3D tissue labeling approach. This solution is comprised of PBS/0.2% Tween  20/Heparin/3% donkey serum/5% DMSO
Caution: For best results, refrigerate after opening.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty