Would you like to log in again?
BD Veritor™ Plus System Rapid, Reliable SARS-CoV-2 (Covid-19) Antigen Testing at the Point of Care, BD
Supplier: BD Diagnostics
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
256082 445006
76423-716KT 391.95 USD InStock
76423-716 76423-718
BD Veritor™ Plus System Rapid, Reliable SARS-CoV-2 (Covid-19) Antigen Testing at the Point of Care, BD
Assays Immunological Assays
The BD Veritor™ Plus System is a handheld device that enables testing for Flu A+B, Group A Strep, and respiratory syncytial virus (RSV) assays. One-button functionality and workflow flexibility allow the healthcare team to confidently use the device. In addition, the system’s fast and accurate results may help improve the patient experience. The BD Veritor™ Plus System streamlines the point-of-care testing experience.
  • Easy operation and 1-button functionality may potentially reduce manual test processing errors
  • Enables intuitive sample processing with prefilled, unitized tubes color-coded by reagent or assay type
  • Displays easy-to-read digital results for SARS-CoV-2 in 15 minutes
  • Records results on secured drive
  • Advanced particle technology enhances sensitivity by using a proprietary process to produce highly stable, modified colloidal metal particles, helping improve test performance
  • Adaptive read technology helps improve specificity to reduce false-positive results by compensating for background and nonspecific binding
Note: The Analyzer part number is 75846-772.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty