Would you like to log in again?
Whatman™ Grade GF 8 Glass Microfiber Filters with Binder, Whatman products (Cytiva)
Supplier: Whatman products (Cytiva)
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Whatman™
10370105 10370111 10370172 10370119 10370172 10370105 10370119 10370111
10034-838PK 178.78 USD InStock
10034-838 10034-836 CA11013-420 CA11013-562 10034-834 CA76423-722 76423-722 CA11013-858
Whatman™ Grade GF 8 Glass Microfiber Filters with Binder, Whatman products (Cytiva)
Filters Glass Fiber Filters
These glass microfibre filters made from borosilicate glass are used for filtration of coarse particles. They offer faster filtration and a higher air flow rate compared to the similar grade GF 9 filters.
  • Inorganic binder for increased strength
  • Fast filtration speed
  • Can be used as a prefilter for dense samples
  • Function as air pollution filters for detecting furans, PCB, and other chemicals
With the added strength of an inorganic binder, grade GF 8 glass microfibre filters serve as air pollution filters by retaining a range of heavy particles found in vapor samples.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty