Would you like to log in again?
Whatman™ Grade GF 10 Glass Microfiber Filters with Binder, Whatman products (Cytiva)
Supplier: Whatman products (Cytiva)
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Whatman™
10370308 10370320 10370319 10370302 10370393 10370392 10370319 10370391 10370320 10370384 10370305 10370394 10370305 10370391 10370393 10370381 10370308 10370302
10034-846PK 147.03 USD InStock
CA89428-196 CA89428-198 CA11013-840 CA11013-542 CA11013-860 CA76333-760 CA11013-464 CA89428-194 CA11013-452 10034-846 10034-854 CA76423-724 10034-852 10034-850 76423-724 76322-120 CA89187-082 10034-848
Whatman™ Grade GF 10 Glass Microfiber Filters with Binder, Whatman products (Cytiva)
Filters Glass Fiber Filters
These glass microfibre filters made from borosilicate glass are strengthened with an organic binder to provide high mechanical stability.
  • Medium filtration speed
  • Can be used as a roll filter in automated applications
These filters can be used as a weighing aid during infrared weighing or for automatic sampling applications. They also come in reel format for use as a roll filter in automatic air filtration units.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty