Would you like to log in again?
Whatman™ Quartz Air Sampling Filter, Grade QM-B, Whatman products (Cytiva)
Supplier: Whatman products (Cytiva)
Whatman™
1852-042 1852-040 1852-040 1852-042
76423-730PK 211 USD InStock
76423-730 CA76423-730 76423-732 CA76423-732
Whatman™ Quartz Air Sampling Filter, Grade QM-B, Whatman products (Cytiva)
Filters Glass Fiber Filters
These high purity QM-B quartz fibre filters are designed for use in specific air monitoring applications.
QM-B filters are similar to QM-A quartz fibre filters, which are recommended more commonly for air testing applications. By comparison, QM-B is approximately twice as heavy, twice as thick and offers slower flow rates. Because of these characteristics, QM-B can be used in air monitoring applications requiring increased strength or durability.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty