Would you like to log in again?
Whatman™ Application-Specific Filter Papers, Grade 1450CV, Whatman products (Cytiva)
Supplier: Whatman products (Cytiva)
Whatman™
10313209 10313209
76423-752PK 7.56 USD InStock
76423-752 CA76423-752
Whatman™ Application-Specific Filter Papers, Grade 1450CV, Whatman products (Cytiva)
Filters Filter Paper Qualitative Filter Paper
Filter paper for identification of undissolved dyes in the textile industry.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty