Would you like to log in again?
Whatman™ Pressure Filtration Devices, Whatman products (Cytiva)
Supplier: Whatman products (Cytiva)
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Whatman™
10452006 10451710 10451610 10451610 10450450 10451710 10450450
10035-072PK 5826.35 USD InStock
10035-072 CA11013-478 CA11013-114 10035-066 CA11013-088 10035-070 76423-824
Whatman™ Pressure Filtration Devices, Whatman products (Cytiva)
Filters Membrane Filter Units
Pressure filtration devices with a sample loading cylinder are suitable for batch filtration of samples from 20 ml. Devices without infusion cylinders are connected inline and are suitable for larger volumes of several liters.
Filtration of liquids and gases is possible, including sterile filtration of serums or clear filtration of media that are difficult to filter, especially those that are highly viscous. Membranes, paper or glass fiber filter discs can be used. Cleaning and changing of filters is completed in a few steps.
All units are equipped with pressure-resistant filter supports. High-quality silicone or PTFE O-rings seal the systems. Please ensure that you use only intact seals for safety reasons. PTFE versions are available in addition to stainless steel devices, for use with corrosive media.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty