Would you like to log in again?
Edge® 48-919 Light-Duty Multi-Purpose Industrial Gloves, Ansell
Supplier: Ansell
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Edge®
48919070 48919060 48919100 48919080 48919090 48919110
76424-286CS 608.53 USD InStock
76424-286 76424-284 76424-288 76424-290 76424-294 76424-292
Edge® 48-919 Light-Duty Multi-Purpose Industrial Gloves, Ansell
Gloves
Cost effective solution for lightweight applications requiring a durable coating and secure grip in oily environments.
  • Excellent abrasion resistance
  • Double layer nitrile coating keeps hands dry and comfortable when handling oily parts and tools
  • Elastic knit-wrist ensures snug fit
  • Applicable in oily environments
Made of a seamless liner, fully dipped into NBR coating. The gloves are fully NBR dipped to provide protection against industrial fluid exposure. The rough palm coating ensures high secure grip in oily environment. Designed for workers who handle small and medium-sized components coated with oil or lubricant. Use primarily for automotive, construction, machinery/equipment, and metal fabrication applications.
Certifications: EN 388:2016. Category II.
Packaging: Gloves are packaged 12 pairs per bag, 12 bags per carton.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty