Would you like to log in again?
Edge® 48-929 Medium-Duty Cut Resistant Industrial Gloves, Ansell
Supplier: Ansell
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Edge®
48929070 48929080 48929060 48929110 48929100 48929090
76424-298CS 1519.6 USD InStock
76424-298 76424-302 76424-300 76424-306 76424-304 76424-308
Edge® 48-929 Medium-Duty Cut Resistant Industrial Gloves, Ansell
Gloves
Cost effective solution for medium weight applications requiring cut resistance, excellent grip, and strong oil-repellence.
  • Excellent abrasion resistance
  • Double layer nitrile coating keeps hands dry and comfortable when handling oily parts and tools
  • Elastic knitwrist ensures snug fit
  • Applicable in oily environments
Made of a seamless liner, fully dipped into NBR coating. The gloves are fully NBR dipped to protect skin from oil contamination. The palm rough NBR coating ensures a highly secure grip in oily environments. Use primarily for aerospace, automotive, glass manufacturing, machinery/equipment, metal fabrication, and transportation applications.
Certifications: EN 388:2016. EN 420:2003 + A1:2009. Category II.
Packaging: Gloves are packaged 12 pairs per bag, 12 bags per carton.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty