Would you like to log in again?
WHEATON® 13 mm Flip-Cap Long Skirt Aluminum Seals, DWK Life Sciences
Supplier: DWK Life Sciences
224200LS-100 224201LS-100
76424-324CS 26.03 USD InStock
76424-324 76424-326
WHEATON® 13 mm Flip-Cap Long Skirt Aluminum Seals, DWK Life Sciences
Closures Chromatography Vial Closures
The longer skirt of the aluminum seal aids in completeness of crimp.
  • Longer Skirt Length
  • Manufactured from lightweight aluminum
  • Two color choices for easy product identification
  • Protects vial from contaminates
  • Can be autoclaved
  • Custom configuration available upon request
  • Lot controlled
There are two key problems with crimping; Incomplete crimp and over crimping. Incomplete crimping can compromise the closure container integrity through ease of removal. However, overcrimping used to overcome incomplete crimping can also compromise the closure container integrity and introduce a number of cosmetic defects like buckling and tears of the skirt. The long skirt seal consists of the same great materials as our other seals, just in a longer skirt length to assure completeness of crimp.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty