Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
ECOMPARATOR EC3000 ASTM CLR 106X57X210MM
Supplier: Lovibond
Ecomparator, EC 3000 ASTM Color, Progressing from Visual to Automatic Colour Measurement, Colour Type: Transparent, Display: Size: 3.5in; Resolution: 320 x 240; Colour: 24 Bit, Interface: USB, Keypad: 3 key tactile membrane, Instrument Housing: Flame Retardant ABS, Portable, Dimensions: 106x57x210mm
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
ECOMPARATOR EC3000 ASTM CLR 106X57X210MM 76424-362 163010 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty