Really versatile general purpose mid-polarity phase for a wide range of applications. Ideal in confirmation analysis.
- Phase composition: Crossbonded Silphenylene - 35% Phenyl - 65% Methyl Polysiloxane
- Long lifetime
Max temperature limits may change depending on the film thickness.