General purpose low-to-mid-polarity phase for a wide range of applications, including (for example): Residual solvents, volatile pollutants.
- Phase composition: Crossbonded 6% cyanopropylphenyl, 94% Methyl polysiloxane
- Long lifetime
Max temperature limits may change depending on the film thickness.