Would you like to log in again?
Avantor® Hichrom HI-BASIC, GC Columns
Supplier: VWR
Avantor® Hichrom
HI38-32-025-30 HI38-32-025-25 HI38-25-015-25
76426-548EA 1847.85 USD InStock
76426-548 76426-546 76426-544
Avantor® Hichrom HI-BASIC, GC Columns
Chromatography Columns
Application specific column for basic compouns (e.g. amines).
Max temperature limits may change depending on the film thickness.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty