Would you like to log in again?
Avantor® Hichrom HI-BIODIESEL 110, GC Columns
Supplier: VWR
Avantor® Hichrom
HI41-32-180-30 HI41-32-100-30
76426-558EA 1801.3 USD InStock
76426-558 76426-560
Avantor® Hichrom HI-BIODIESEL 110, GC Columns
Chromatography Columns
Application specifc column for free and total glycerine (phase stable up to 380 °C) and for FAMEs in biodiesel analysis.
Max temperature limits may change depending on the film thickness.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty