Would you like to log in again?
Avantor® Hichrom HI-DAI 1, GC Columns
Supplier: VWR
Avantor® Hichrom
HI42-32-500-50 HI42-32-500-30 HI42-25-250-30 HI42-25-250-50 HI42-32-300-50 HI42-32-300-30
76426-568EA 2900.66 USD InStock
76426-568 76426-566 76426-564 76426-562 76426-572 76426-570
Avantor® Hichrom HI-DAI 1, GC Columns
Chromatography Columns
Application specifc column for direct introduction of aqueous samples, thus minimizing sample preparation.
Max temperature limits may change depending on the film thickness.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty