Very popular nitroterephthalic-acid-modified polyethylene glycol (PEG) column of high polarity for the analysis of free fatty acids, phenols, alcohols, glycols, with extended temperature range.
- Phase composition: Extended temperature crossbonded nitroterephthalic-acid-modified polyethylene glycol (PEG)
- Long lifetime
Max temperature limits may change depending on the film thickness.