Would you like to log in again?
Avantor® Hichrom HI-LAP, GC Columns
Supplier: VWR
Avantor® Hichrom
HI46-32-010-25 HI46-25-010-25
76427-120EA 364.1 USD InStock
76427-120 76427-122
Avantor® Hichrom HI-LAP, GC Columns
Chromatography Columns
Application specific column for lipids, sterols and triglycerides analysis.
Maximum temperature limits may change depending on the film thickness.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty