Would you like to log in again?
Avantor® Hichrom HI-DEX DAC Beta, GC Columns
Supplier: VWR
Avantor® Hichrom
HI66-25-025-30 HI78-10-010-5 HI66-25-015-30 HI78-10-010-10 HI66-25-025-50 HI66-25-015-25 HI66-25-025-25 HI66-25-015-15 HI66-25-025-15 HI66-25-015-60 HI66-25-025-60 HI66-25-015-50
76429-894EA 403.75 USD InStock
76429-894 76429-908 76429-898 76429-910 76429-896 76429-902 76430-062 76429-900 76430-064 76429-912 76429-906 76429-904
Avantor® Hichrom HI-DEX DAC Beta, GC Columns
Chromatography Columns
Useful for the analysis of chiral compounds in fragrances, pesticides and pharmaceuticals in which the determination of the ratio of enantiomers in a sample is necessary.
Max temperature limits may change depending on the film thickness.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty