Would you like to log in again?
Avantor® Hichrom HI-DEX DAC Gamma, GC Columns
Supplier: VWR
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Avantor® Hichrom
HI67-25-025-60 HI67-25-015-15 HI79-10-010-5 HI79-10-010-10 HI67-25-015-60 HI67-25-025-30 HI67-25-025-15 HI67-25-015-30 HI67-25-015-25 HI67-25-015-50 HI67-25-025-50 HI67-25-025-25
76429-920EA 3590.22 USD InStock
76429-920 76429-932 76429-930 76429-924 76429-914 76429-922 76429-928 76430-066 76429-918 76429-926 76430-068 76429-916
Avantor® Hichrom HI-DEX DAC Gamma, GC Columns
Chromatography Columns
Useful for the analysis of chiral compounds in fragrances, pesticides and pharmaceuticals in which the determination of the ratio of enantiomers in a sample is necessary.
Maximum temperature limits may change depending on the film thickness.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty